Assay Details
| Gene Symbol: vdac3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011249.2 | NP_001011249.1 | 1 - 2 | 75 | 66 | |
| XM_012958925.2 | XP_012814379.1 | 2 - 3 | 171 | 66 | ||
| XM_012958926.2 | XP_012814380.1 | 2 - 3 | 171 | 66 | ||
| XM_012958927.2 | XP_012814381.1 | 2 - 3 | 95 | 66 | ||
| XM_012958929.1 | XP_012814383.1 | 2 - 3 | 151 | 66 | ||
| XM_012958930.2 | XP_012814384.1 | 2 - 3 | 157 | 66 | ||
| XM_012958931.1 | XP_012814385.1 | 2 - 3 | 151 | 66 | ||
| XM_012958932.2 | XP_012814386.1 | 2 - 3 | 155 | 66 | ||
| GenBank mRNA | BC087828.1 | - | 1 - 2 | 76 | 66 | |
More Information
Set Membership: |
|

vdac3
Genomic Map