Assay Details
Gene Symbol: gsto1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011256.1 | NP_001011256.1 | 1 - 2 | 74 | 70 | |
XM_012966511.2 | XP_012821965.1 | 2 - 3 | 173 | 70 | ||
GenBank mRNA | BC087558.2 | - | 1 - 2 | 74 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
