Assay Details
| Gene Symbol: gamt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203565.1 | NP_988896.1 | 4 - 5 | 467 | 67 | |
| GenBank mRNA | BC059738.1 | - | 4 - 5 | 467 | 67 | |
| CR760866.2 | - | 4 - 5 | 474 | 67 | ||
More Information
Set Membership: |
|

gamt
Genomic Map