Assay Details
Gene Symbol: gamt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203565.1 | NP_988896.1 | 5 - 6 | 580 | 92 | |
GenBank mRNA | BC059738.1 | - | 5 - 6 | 580 | 92 | |
CR760866.2 | - | 5 - 6 | 587 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
