Assay Details
| Gene Symbol: yaf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203566.1 | NP_988897.1 | 2 - 3 | 251 | 59 | |
| GenBank mRNA | BC059734.1 | - | 2 - 3 | 251 | 59 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

yaf2| Gene Symbol: yaf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203566.1 | NP_988897.1 | 2 - 3 | 251 | 59 | |
| GenBank mRNA | BC059734.1 | - | 2 - 3 | 251 | 59 | |
Set Membership: |
|