Assay Details
Gene Symbol: emc10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203571.1 | NP_988902.1 | 1 - 2 | 131 | 82 | |
XM_012956621.2 | XP_012812075.1 | 1 - 2 | 218 | 82 | ||
GenBank mRNA | BC061625.1 | - | 1 - 2 | 131 | 82 | |
CR760224.2 | - | 1 - 2 | 136 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
