Assay Details
Gene Symbol: dhps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203575.1 | NP_988906.1 | 1 - 2 | 222 | 58 | |
GenBank mRNA | BC061622.1 | - | 1 - 2 | 222 | 58 | |
CR760799.2 | - | 1 - 2 | 230 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
