Assay Details
| Gene Symbol: slc25a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203578.1 | NP_988909.1 | 1 - 2 | 169 | 97 | |
| GenBank mRNA | BC061600.1 | - | 1 - 2 | 169 | 97 | |
| CR761556.2 | - | 1 - 2 | 189 | 97 | ||
More Information
Set Membership: |
|

slc25a4
Genomic Map