Assay Details
Gene Symbol: akirin2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203583.1 | NP_988914.1 | 3 - 4 | 708 | 88 | |
GenBank mRNA | BC061612.1 | - | 3 - 4 | 708 | 88 | |
CR760919.2 | - | 3 - 4 | 520 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
