Assay Details
Gene Symbol: ap3s2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011184.2 | NP_001011184.1 | 3 - 4 | 307 | 90 | |
XM_012958920.2 | XP_012814374.1 | 3 - 4 | 507 | 90 | ||
GenBank mRNA | BC086503.2 | - | 3 - 4 | 307 | 90 | |
CR761527.2 | - | 3 - 4 | 321 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
