Assay Details
Gene Symbol: mogat2.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011301.1 | NP_001011301.1 | 1 - 2 | 268 | 59 | |
GenBank mRNA | BC088019.1 | - | 1 - 2 | 268 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
