Assay Details
Gene Symbol: mogat2.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011301.1 | NP_001011301.1 | 3 - 4 | 638 | 65 | |
GenBank mRNA | BC088019.1 | - | 3 - 4 | 638 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
