Assay Details
Gene Symbol: hsd17b11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011304.1 | NP_001011304.1 | 6 - 7 | 874 | 67 | |
GenBank mRNA | BC088026.1 | - | 6 - 7 | 874 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
