Assay Details
Gene Symbol: pomc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011318.1 | NP_001011318.1 | 2 - 3 | 184 | 66 | |
GenBank mRNA | BC088054.1 | - | 2 - 3 | 184 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
