Assay Details
| Gene Symbol: cdca7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011378.1 | NP_001011378.1 | 2 - 3 | 149 | 82 | |
| GenBank mRNA | BC088559.1 | - | 2 - 3 | 149 | 82 | |
| CR855398.2 | - | 2 - 3 | 143 | 82 | ||
More Information
Set Membership: |
|

cdca7
Genomic Map