Assay Details
Gene Symbol: emc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011398.1 | NP_001011398.1 | 2 - 3 | 140 | 65 | |
GenBank mRNA | BC088602.1 | - | 2 - 3 | 140 | 65 | |
CR848509.2 | - | 2 - 3 | 146 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
