Assay Details
Gene Symbol: serinc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011399.1 | NP_001011399.1 | 3 - 4 | 518 | 59 | |
XM_018093869.1 | XP_017949358.1 | 3 - 4 | 478 | 59 | ||
GenBank mRNA | BC088606.1 | - | 3 - 4 | 518 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
