Assay Details
| Gene Symbol: mgmt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011404.2 | NP_001011404.2 | 5 - 6 | 519 | 54 | |
| NM_001244868.1 | NP_001231797.1 | 5 - 6 | 514 | 54 | ||
| XM_018095366.1 | XP_017950855.1 | 5 - 6 | 1310 | 54 | ||
| XM_018095368.1 | XP_017950857.1 | 6 - 7 | 735 | 54 | ||
| XM_018095369.1 | XP_017950858.1 | 4 - 5 | 528 | 54 | ||
| XM_018095370.1 | XP_017950859.1 | 4 - 5 | 507 | 54 | ||
| XM_018095371.1 | XP_017950860.1 | 4 - 5 | 492 | 54 | ||
| GenBank mRNA | BC088767.1 | - | 5 - 6 | 513 | 54 | |
| BX741093.1 | - | 4 - 5 | 482 | 54 | ||
| DN095842.1 | - | 5 - 6 | 514 | 54 | ||
More Information
Set Membership: |
|

mgmt
Genomic Map