Assay Details
| Gene Symbol: angptl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011408.1 | NP_001011408.1 | 4 - 5 | 845 | 84 | |
| GenBank mRNA | BC088778.1 | - | 4 - 5 | 845 | 84 | |
| CT030514.1 | - | 4 - 5 | 859 | 84 | ||
More Information
Set Membership: |
|

angptl3
Genomic Map