Assay Details
| Gene Symbol: rpl32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011414.1 | NP_001011414.1 | 3 - 4 | 306 | 86 | |
| GenBank mRNA | BC087976.1 | - | 3 - 4 | 306 | 86 | |
| CU025172.1 | - | 3 - 4 | 298 | 86 | ||
More Information
Set Membership: |
|

rpl32
Genomic Map