Assay Details
| Gene Symbol: uox | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011416.1 | NP_001011416.1 | 1 - 2 | 47 | 62 | |
| GenBank mRNA | BC087991.1 | - | 1 - 2 | 47 | 62 | |
More Information
Set Membership: |
|

uox
Genomic Map