Assay Details
| Gene Symbol: eef1a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011418.1 | NP_001011418.1 | 3 - 4 | 655 | 69 | |
| GenBank mRNA | BC088010.1 | - | 3 - 4 | 655 | 69 | |
More Information
Set Membership: |
|

eef1a2
Genomic Map