Assay Details
Gene Symbol: cdk5rap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011442.1 | NP_001011442.1 | 7 - 8 | 656 | 72 | |
GenBank mRNA | BC088507.1 | - | 7 - 8 | 656 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
