Assay Details
| Gene Symbol: eif2s3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011456.1 | NP_001011456.1 | 2 - 3 | 140 | 59 | |
| XM_018091216.1 | XP_017946705.1 | 2 - 3 | 383 | 59 | ||
| GenBank mRNA | BC088563.1 | - | 2 - 3 | 140 | 59 | |
More Information
Set Membership: |
|

eif2s3
Genomic Map