Assay Details
Gene Symbol: sike1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011474.1 | NP_001011474.1 | 1 - 2 | 187 | 96 | |
XM_012961449.2 | XP_012816903.1 | 1 - 2 | 1910 | 96 | ||
XM_012961451.1 | XP_012816905.1 | 2 - 3 | 379 | 96 | ||
GenBank mRNA | BC088764.1 | - | 1 - 2 | 187 | 96 | |
CR848255.2 | - | 1 - 2 | 185 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
