Assay Details
Gene Symbol: dnajc25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011497.2 | NP_001011497.1 | 1 - 2 | 385 | 76 | |
GenBank mRNA | BC088889.1 | - | 1 - 2 | 386 | 76 | |
CT027859.1 | - | 1 - 2 | 407 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
