Assay Details
Gene Symbol: casc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012652.1 | NP_001012670.1 | 1 - 2 | 184 | 62 | |
XM_012971726.2 | XP_012827180.1 | 1 - 2 | 62 | |||
XM_012971728.2 | XP_012827182.1 | 1 - 2 | 62 | |||
XM_018089430.1 | XP_017944919.1 | 1 - 2 | 62 | |||
GenBank mRNA | CR855779.2 | - | 1 - 2 | 273 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
