Assay Details
Gene Symbol: stk25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015684.1 | NP_001015684.1 | 7 - 8 | 1113 | 70 | |
XM_012970374.2 | XP_012825828.1 | 8 - 9 | 1101 | 70 | ||
XM_012970375.2 | XP_012825829.1 | 8 - 9 | 1083 | 70 | ||
XM_018093870.1 | XP_017949359.1 | 8 - 9 | 1057 | 70 | ||
GenBank mRNA | BC089072.1 | - | 7 - 8 | 1113 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
