Assay Details
| Gene Symbol: p2ry13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015696.1 | NP_001015696.1 | 1 - 2 | 232 | 82 | |
| GenBank mRNA | BC089239.1 | - | 1 - 2 | 232 | 82 | |
More Information
Set Membership: |
|

p2ry13
Genomic Map