Assay Details
| Gene Symbol: csnk1g2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015725.1 | NP_001015725.1 | 6 - 7 | 1235 | 67 | |
| XM_012961607.2 | XP_012817061.1 | 8 - 9 | 1407 | 67 | ||
| GenBank mRNA | BC089657.1 | - | 6 - 7 | 1235 | 67 | |
More Information
Set Membership: |
|

csnk1g2
Genomic Map