Assay Details
Gene Symbol: ctsl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015747.1 | NP_001015747.1 | 7 - 8 | 1118 | 68 | |
XM_012966538.2 | XP_012821992.1 | 7 - 8 | 1022 | 68 | ||
GenBank mRNA | BC089683.1 | - | 7 - 8 | 1118 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
