Assay Details
Gene Symbol: proc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015759.1 | NP_001015759.1 | 4 - 5 | 348 | 64 | |
GenBank mRNA | BC089695.1 | - | 4 - 5 | 348 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
