Assay Details
| Gene Symbol: cxcl12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015764.1 | NP_001015764.1 | 2 - 3 | 206 | 64 | |
| GenBank mRNA | BC089703.1 | - | 2 - 3 | 206 | 64 | |
More Information
Set Membership: |
|

cxcl12
Genomic Map