Assay Details
| Gene Symbol: tmed4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015779.1 | NP_001015779.1 | 2 - 3 | 292 | 68 | |
| GenBank mRNA | BC089720.1 | - | 2 - 3 | 292 | 68 | |
| CT030280.1 | - | 2 - 3 | 297 | 68 | ||
More Information
Set Membership: |
|

tmed4
Genomic Map