Assay Details
Gene Symbol: c2orf40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015782.1 | NP_001015782.2 | 4 - 5 | 463 | 82 | |
GenBank mRNA | BC089724.1 | - | 4 - 5 | 463 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
