Assay Details
| Gene Symbol: eef2.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015785.1 | NP_001015785.1 | 2 - 3 | 468 | 59 | |
| GenBank mRNA | BC089730.1 | - | 2 - 3 | 468 | 59 | |
More Information
Set Membership: |
|

eef2.2
Genomic Map