Assay Details
| Gene Symbol: prss1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015792.1 | NP_001015792.1 | 1 - 2 | 49 | 85 | |
| GenBank mRNA | BC089741.1 | - | 1 - 2 | 49 | 85 | |
More Information
Set Membership: |
|

prss1
Genomic Map