Assay Details
| Gene Symbol: acyp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015794.1 | NP_001015794.1 | 3 - 4 | 208 | 64 | |
| GenBank mRNA | BC089743.1 | - | 3 - 4 | 208 | 64 | |
More Information
Set Membership: |
|

acyp2
Genomic Map