Assay Details
| Gene Symbol: cd37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015801.2 | NP_001015801.2 | 8 - 9 | 831 | 64 | |
| GenBank mRNA | BC089757.1 | - | 8 - 9 | 831 | 64 | |
More Information
Set Membership: |
|

cd37
Genomic Map