Assay Details
| Gene Symbol: afg3l1p | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015906.3 | NP_001015906.1 | 1 - 2 | 142 | 79 | |
| GenBank mRNA | BC170788.1 | - | 1 - 2 | 141 | 79 | |
| BC171124.1 | - | 1 - 2 | 141 | 79 | ||
| CR926435.2 | - | 1 - 2 | 142 | 79 | ||
More Information
Set Membership: |
|

afg3l1p
Genomic Map