Assay Details
Gene Symbol: afg3l1p | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015906.3 | NP_001015906.1 | 5 - 6 | 547 | 72 | |
GenBank mRNA | BC170788.1 | - | 5 - 6 | 546 | 72 | |
BC171124.1 | - | 5 - 6 | 546 | 72 | ||
CR926435.2 | - | 5 - 6 | 547 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
