Assay Details
Gene Symbol: ocln | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015948.1 | NP_001015948.1 | 6 - 7 | 1377 | 67 | |
GenBank mRNA | BC123964.1 | - | 6 - 7 | 1370 | 67 | |
CR761445.2 | - | 6 - 7 | 1377 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
