Assay Details
| Gene Symbol: trappc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015966.2 | NP_001015966.1 | 1 - 2 | 95 | 71 | |
| GenBank mRNA | BC091013.1 | - | 1 - 2 | 117 | 71 | |
| CR848424.2 | - | 1 - 2 | 95 | 71 | ||
More Information
Set Membership: |
|

trappc3
Genomic Map