Assay Details
| Gene Symbol: mrps36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015969.2 | NP_001015969.1 | 3 - 4 | 328 | 93 | |
| XM_012962955.2 | XP_012818409.1 | 3 - 4 | 429 | 93 | ||
| GenBank mRNA | BC118871.1 | - | 3 - 4 | 328 | 93 | |
| CR848415.2 | - | 3 - 4 | 328 | 93 | ||
More Information
Set Membership: |
|

mrps36
Genomic Map