Assay Details
Gene Symbol: sh3rf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015973.2 | NP_001015973.1 | 2 - 3 | 722 | 81 | |
XM_012961866.2 | XP_012817320.1 | 2 - 3 | 1004 | 81 | ||
XM_012961871.2 | XP_012817325.1 | 2 - 3 | 1033 | 81 | ||
XM_012961878.2 | XP_012817332.1 | 2 - 3 | 1002 | 81 | ||
XM_018093029.1 | XP_017948518.1 | 2 - 3 | 1041 | 81 | ||
XM_018093030.1 | XP_017948519.1 | 2 - 3 | 1037 | 81 | ||
GenBank mRNA | BC167346.1 | - | 2 - 3 | 780 | 81 | |
BC169143.1 | - | 1 - 2 | 459 | 81 | ||
BC171011.1 | - | 2 - 3 | 580 | 81 | ||
CR848380.2 | - | 2 - 3 | 722 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
