Assay Details
| Gene Symbol: asmtl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016011.2 | NP_001016011.1 | 10 - 11 | 1326 | 89 | |
| GenBank mRNA | BC121542.1 | - | 9 - 10 | 1294 | 89 | |
| CR848221.2 | - | 10 - 11 | 1326 | 89 | ||
More Information
Set Membership: |
|

asmtl
Genomic Map