Assay Details
Gene Symbol: ube2l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016063.2 | NP_001016063.1 | 1 - 2 | 39 | 83 | |
GenBank mRNA | BC091621.1 | - | 1 - 2 | 39 | 83 | |
CR762273.2 | - | 1 - 2 | 39 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
