Assay Details
| Gene Symbol: zc3h15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016105.2 | NP_001016105.1 | 7 - 8 | 968 | 71 | |
| GenBank mRNA | BC135295.1 | - | 7 - 8 | 889 | 71 | |
| CR762127.2 | - | 7 - 8 | 968 | 71 | ||
More Information
Set Membership: |
|

zc3h15
Genomic Map