Assay Details
| Gene Symbol: eri1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016146.2 | NP_001016146.1 | 2 - 3 | 332 | 79 | |
| GenBank mRNA | BC155977.1 | - | 2 - 3 | 594 | 79 | |
| BC155987.1 | - | 2 - 3 | 594 | 79 | ||
| CR761994.2 | - | 2 - 3 | 332 | 79 | ||
More Information
Set Membership: |
|

eri1
Genomic Map