Assay Details
Gene Symbol: cyb5d2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016154.1 | NP_001016154.1 | 3 - 4 | 626 | 82 | |
GenBank mRNA | BC161279.1 | - | 3 - 4 | 610 | 82 | |
BC170615.1 | - | 3 - 4 | 622 | 82 | ||
BC170621.1 | - | 3 - 4 | 622 | 82 | ||
CR761951.1 | - | 3 - 4 | 626 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
